Wood Funnel Support, Adjustable Bracket, Bennett Wood
Supplier: Bennett Wood Specialties
2608
30575-001EA 369.87 USD
30575-001
Wood Funnel Support, Adjustable Bracket, Bennett Wood
Support Stands
Holds six funnels
  • Without glassware
  • With bracket adjustable on each end
Shelf is adjustable over a height of 127 mm (5"). Slotted funnel openings spaced 133 mm (51/4") center to center.