SP Bel-Art Rotary Separatory Funnel Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H18964-0000
30534-900EA 319.4 USD
30534-900
SP Bel-Art Rotary Separatory Funnel Rack, Bel-Art Products, a part of SP
Support Stands
Rack consists of a rectangular base, screw-in column, and a support disc held in place by a screw clamp.
  • Fully adjustable support disc
  • Open front for easy removal and placement of separatory funnels
  • Insert clips accommodate various sizes of funnels
The support disc is fully adjustable for different heights up to 53.3 cm (21"). Designed to accommodate a maximum of six 500 mL or 1000 mL funnels.
Delivery information: Funnel rack includes 18 insert clips used to accommodate different sized separatory funnels. Shipped unassembled.