PYREX® Separatory Squibb Funnel, with PYREX® Stopper and Rotaflo Stopcock, Corning
Supplier: Corning
6302-125
30356-460CS 285.84 USD
30356-460 30356-449 30356-482
PYREX® Separatory Squibb Funnel, with PYREX® Stopper and Rotaflo Stopcock, Corning
Funnels Separating Funnels
Made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
  • Stem Ext.Ø. is 10 mm
These separatory funnels have Rotaflo® stopcocks and strong standard taper glass stoppers.