PYREX® Globe-Shaped Separatory Funnels with [ST] Glass Stopper and [ST] Stopcock, Corning
Supplier: Corning
6340-1L
30341-102CS 384.98 USD
30341-102
PYREX® Globe-Shaped Separatory Funnels with [ST] Glass Stopper and [ST] Stopcock, Corning
Funnels Separating Funnels
PYREX®, borosilicate glass, with glass stopper.
These separatory funnels are made from molded blanks which are tapered toward the stopcock to facilitate the separation of liquids. The sturdy design makes it suitable for rack work.