Analytical Funnel, Polypropylene, Dynalon
Supplier: Dynalon
241665
30286-500CS 785.91 USD
30286-500
Analytical Funnel, Polypropylene, Dynalon
Funnels Filtration Funnels Analytical Funnels
Made of autoclavable polypropylene, these funnels have long a narrow bore that works best for liquids