SP Bel-Art Draft Checker™ Airflow Detection Powder, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F19939-0000
30181-800EA 48.06 USD
30181-800
SP Bel-Art Draft Checker™ Airflow Detection Powder, Bel-Art Products, a part of SP
Spill Control Materials
A quick, easy way to test the safety of fume hood exhaust before use.
  • Plastic bottle easily dispenses ultra-fine silica powder
  • Inert
  • Non-toxic
  • Non-flammable
Just a squeeze or two from the plastic bottle dispenses a cloud of ultra-fine, lightweight silica powder, which remains suspended long enough to visually check whether or not the fume hood exhaust is functioning properly.
It is also useful for detecting drafts, gas leaks, and air currents in rooms, ducts, and exhaust systems. The amorphous silica powder is inert, non-toxic, and non-flammable.
Packaging: Packaged in a 1.5 g bottle.