SP Bel-Art Large Molded Fume Hood, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H50000-0003
30181-775EA 1511.26 USD
30181-775
SP Bel-Art Large Molded Fume Hood, Bel-Art Products, a part of SP
Fume Hoods and Enclosures Chemical Fume Hoods
This fume hood features a one-piece shell molded of polyethylene plastic with a clear acrylic sash that slides on inside tracks.
  • One-piece design
  • Polyethylene plastic construction
  • Clear acrylic sash slides on inside tracks
  • Sash facilitates continuous airflow
It can be set in an open, closed or intermediate position. The front opening is 88 x 36 cmH (34½ x 14³/₁₆"). When in the closed position, the sash remains open slightly to facilitate continuous air flow. Stack O.D. is 15 cm (6").
Ordering information: An exhaust system capable of drawing 350 cfm (free air) is recommended.