PS-90 SensaTemp® Production Soldering Iron, PACE®
Supplier: Pace
6993-0199-P1
30138-506EA 562.74 USD
30138-506
PS-90 SensaTemp® Production Soldering Iron, PACE®
Soldering Supplies Soldering Irons
For heavy-duty soldering and general-purpose and SMT rework operations
Provides high thermal capacity and flexibility. Handpiece features PermaGround™ tip grounding system that ensures component safety.
Ordering information: Comes with tip and tool stand. For soldering tips, see 30138-116.