Blow Guns, 600, Rubber Tip, Coilhose Pneumatics
Supplier: Safety & Industrial Supplies
166-601
300087-790EA 37.47 USD
300087-790
Blow Guns, 600, Rubber Tip, Coilhose Pneumatics
Pneumatic Tools
Thumb-lever style blowgun with an ergonomic shape.
  • Plated finish provides easy cleaning
  • Tough die-cast zinc body
  • 32 to 175 °F operating temperature range