Strion® LED HP Flashlights, Streamlight®
Supplier: Safety & Industrial Supplies
683-74502
300085-078EA 334.87 USD
300085-078
Strion® LED HP Flashlights, Streamlight®
Lights Flashlights
Includes 2 charger/holders, AC and DC charge cords, and battery.
  • Water resistant, machined aluminum body
  • High powered, rechargeable LED flashlight featuring a C4 LED
  • Lithium-ion battery
  • 50000 hour lamp life
  • 500 charge cycles
  • High, medium, low, or strobe modes
  • One 3.75 V battery