Series 575 Slim Simple Design Digimatic Indicators, Mitutoyo
Supplier: Safety & Industrial Supplies
504-575-123
300082-228EA 289.55 USD
300082-228
Series 575 Slim Simple Design Digimatic Indicators, Mitutoyo
Measuring and Leveling Tools
Ideal for installation into measuring devices because of it's compact design and long battery life.
  • Unlimited response speed eliminates over-speed errors
  • Digital LCD
  • SPC data output
  • Stem mounting
Employing the ABSOLUTE linear encoder, the ID-U always displays the spindle "Absolute Position" from the origin at power-on.
Certifications: IP42 rating