Digital Multimeters, Milwaukee® Electric Tools
Supplier: Safety & Industrial Supplies
495-2217-20
300081-878EA 291.84 USD
300081-878
Digital Multimeters, Milwaukee® Electric Tools
Electrical Meters Digital Multimeters
Simplified user interface feature set specifically for the electrician and HVAC technician.
  • Rugged overmolding for durability
  • Overmolded dial for one handed use
  • Slide rail for mounting voltage detector or magnet accessories
  • Functions include continuity, current, resistance, voltage, frequency
Ordering information: Includes instructions, test leads, multimeter, and thermocouple.