Mini-Triggertorch™ Microtorch Kits, Master Appliance
Supplier: Safety & Industrial Supplies
467-MT-70
300081-420EA 28.09 USD
300081-420 300081-422
Mini-Triggertorch™ Microtorch Kits, Master Appliance
Heaters Hot Air Dryers
Built-in self-igniting system with safety lock that ensures the switch is not turned on when not in use. Contains butane gas.
  • 'Hands-free' slide lock for continuous use and instant off operation
  • Durable self-contained, refillable plastic fuel tank
  • Gas adjustment wheel provides adjustable, long blue torch flame
  • Incorporates 'trigger' switch with 'pulldown' lock to meet Consumer Products Safety Commission (CPSC) Child Safety Requirements
  • Self-igniting piezo electric ignition is easy to start and eliminates the need for lighter or matches
Unique shutter lever to easily extinguish ignition flame if using optional soldering or hot air tip attachments.
Ordering information: The promo pack includes MT-11 Microtorch and MT-70 Mini-Triggertorch.