Advantage® 1000 RCA Gas Masks, MSA
Supplier: Safety & Industrial Supplies
454-813860
300081-134EA 787.81 USD
300081-134
Advantage® 1000 RCA Gas Masks, MSA
Respirators Reusable Respirators Full Face Respirators
This provides a standard mechanical speaking diaphragm, canister and ID tag.
  • Flexible, 1 piece polyurethane lens with wide field of vision
  • Constructed of super-soft Hycar™ rubber
  • Dual-canister mount (bayonet mount) allows weapon sighting from either shoulder
  • Fully elastic, 6 point head harness promotes easy on-off adjustment, no hair pulling
  • With suspension strap
The standard nosecup helps eliminate lens fogging.