Full Facepiece Respirator Cover Lenses, MSA
Supplier: Safety & Industrial Supplies
454-456975
300081-044PK 138.73 USD
300081-044
Full Facepiece Respirator Cover Lenses, MSA
Respirators Respiratory Systems
Full facepiece cover lenses protect the facepiece lens from scratches during storage, handling, and use.
  • Clear, flexible lens cover
  • Easy to install and replace
  • For use with Ultravue Masks