Heavy-Duty SCBA Wall Cases, for Single SCBA, Allegro®
Supplier: Safety & Industrial Supplies
037-4150
300075-252EA 706.21 USD
300075-252
Heavy-Duty SCBA Wall Cases, for Single SCBA, Allegro®
Boxes Dispenser Boxes
Built for simple, fast installation, this wall case is also recyclable and built from Linear Low Density Polyethylene (LLDPE).
This low profile wall mounted case provides protection from dust, moisture, and damaging plant environments. Viewing windows allow easy visibility of the SCBA gauges and internal bracket and straps keep the SCBA secure. Includes universal wall bracket.