High Volume Manual Pumps, Lincoln Industrial
Supplier: Safety & Industrial Supplies
438-1326
300072-256EA 443.75 USD
300072-256
High Volume Manual Pumps, Lincoln Industrial
Pumps Drum/Transfer Pumps
High volume barrel pump with lever action.
  • Neoprene seals resist wear from abrasive fluids
  • Double acting diaphragm
  • Hose with discharge nozzle assures long reach