Wall Mounted Eye Wash/Drench Hose Units, Guardian
Supplier: Safety & Industrial Supplies
333-G5026
300068-338EA 534.13 USD
300068-338
Wall Mounted Eye Wash/Drench Hose Units, Guardian
First Aid Supplies Eyewash Stations and Eyewash Bottles
Two GS-Plus™ spray heads mounted side-by-side, each head has a 'flip top' dust cover, internal flow control and filter to remove impurities from the water flow.
  • Valve has replaceable stainless steel seat for exceptional durability
  • Swivel-type inlet
  • Forged brass squeeze valve activated by stainless steel lever handle
Pressure 300 psi max.
Certifications: ANSI-compliant identification sign.