Counter Mount Eyewash Drench Hose Units, Guardian
Supplier: Safety & Industrial Supplies
333-G5022
300068-334EA 570.14 USD
300068-334
Counter Mount Eyewash Drench Hose Units, Guardian
First Aid Supplies Eyewash Stations and Eyewash Bottles
ANSI-compliant identification sign.
  • Swivel-type inlet
  • Valve has replaceable stainless steel seat for exceptional durability
  • Forged brass squeeze valve activated by stainless steel lever handle
  • Two GS-Plus™ spray heads mounted side-by-side, each head has a 'flip top' dust cover, internal flow control and filter to remove impurities from the water flow
Pressure 300 psi max.