Wall Mounted Eye Washes, Guardian
Supplier: Safety & Industrial Supplies
333-G1814
300068-314EA 490.61 USD
300068-314
Wall Mounted Eye Washes, Guardian
First Aid Supplies Eyewash Stations and Eyewash Bottles
Two GS-Plus™ spray-type spray heads deliver a flood of water for rinsing eyes.
  • ¹/₂" IPS chrome plated brass stay-open ball valve
  • Outlet size 1¼" NPT(F)
Heavy duty cast aluminum wall bracket with corrosion resistant powder coated finish.
Certifications: ANSI-compliant identification sign.