EyeSafe™ Faucet-Mounted Eye Washes, Guardian
Supplier: Safety & Industrial Supplies
333-G1100
300068-296EA 251.85 USD
300068-296
EyeSafe™ Faucet-Mounted Eye Washes, Guardian
First Aid Supplies Eyewash Stations and Eyewash Bottles
Converts any faucet into an emergency eye wash station without interfering with normal faucet operation.
  • Adjustable aerated outlet heads
  • Float off dust covers to protect outlet heads
  • Forged brass diverter valve
  • Body has ⁵⁵/₆₄"-27 female thread
  • Furnished with removable aerator on bottom
Pull knob to activate eye wash; water pressure holds eye wash in operation, leaving user's hands free.