Diaphragm Tilt Valve Balloon Inflators, Western Enterprises
Supplier: Safety & Industrial Supplies
312-RPB-5HG
300066-960EA 163.72 USD
300066-960
Diaphragm Tilt Valve Balloon Inflators, Western Enterprises
Welding Supplies
  • Easily adjusted to increase outlet pressures to 125 psi
  • Equipped with an internal safety relief device
  • Includes filters to protect against contaminant and dirt damage
  • Ideal for use with various latex sizing machines