Flow Gauge Regulators, Western Enterprises
Supplier: Safety & Industrial Supplies
312-REB-4-FG
300066-940EA 289.08 USD
300066-940
Flow Gauge Regulators, Western Enterprises
Welding Supplies
  • Dual scale flow gauge calibrated for Argon/Carbon Dioxide services
  • Durable Neoprene diaphragm maintains consistent flow rates
  • Protective inlet filtering system extends regulator life
  • KEL-F® seat assembly