Flow Gauge Regulators, Argon, Western Enterprises
Supplier: Safety & Industrial Supplies
312-REB-3-FG
300066-934EA 259.55 USD
300066-934 300066-936
Flow Gauge Regulators, Argon, Western Enterprises
Welding Supplies
Dual scale flow gauge calibrated for Argon/Carbon Dioxide services.
  • Durable Neoprene diaphragm maintains consistent flow rates
  • Protective inlet filtering system extends regulator life
  • KEL-F® seat assembly
Regulator 300066-936 includes a 6' Neoprene hose with fittings.
Certifications: UL Listed.