Safemate Quick Connect Sets with Flash Arrestors, Western Enterprises
Supplier: Safety & Industrial Supplies
312-QDF10 312-QDF30
300066-926EA 144.69 USD
300066-926 300066-928
Safemate Quick Connect Sets with Flash Arrestors, Western Enterprises
Welding Supplies
  • Color-coded for gas service to provide added safety
  • Internal check valves designed to prevent reverse flow and accidental mixing of oxygen and fuel gases
  • Torch and regulator models provide downstream and upstream protection
  • 10" maximum cutting capacity