Flashback Arrestor Assemblies, Western Enterprises
Supplier: Safety & Industrial Supplies
312-FA-510CV
300066-810EA 208.29 USD
300066-810
Flashback Arrestor Assemblies, Western Enterprises
Welding Supplies
Designed for installation on acetylene cylinders to prevent flashbacks into the cylinder
  • Extinguish flashes or flames before they reach the cylinder
  • Rated flow capacity is 40 SCFH with a 5 psig pressure drop
  • Includes check valve
Flashback Arrestor Assemblies can be used in any lead not exceeding 3/8" pipe in size which connects an acetylene cylinder to a regulator, manifold, or other equipment.