Flashback Arrestor Bodies, Western Enterprises
Supplier: Safety & Industrial Supplies
312-FA-2
300066-806EA 147.32 USD
300066-806
Flashback Arrestor Bodies, Western Enterprises
Welding Supplies
Extinguish flashes or flames before they reach the cylinder
  • Rated flow capacity is 40 SCFH with a 5 psig pressure drop
  • Use with flash adapter body compatible with 1/4" NPT
Flashback Arrestor Bodies can be used in any lead not exceeding 3/8" pipe in size which connects an acetylene cylinder to a regulator, manifold, or other equipment.