Non-Corrosive Gas Flow Valves, Western Enterprises
Supplier: Safety & Industrial Supplies
312-205 312-206 312-208 312-211 312-211-E 312-218 312-215-AR 312-208-F 312-207 312-215-FR 312-218-D 312-217
300007-814EA 30.73 USD
300007-814 300007-815 300007-816 300007-817 300066-292 300091-794 300066-284 300007-813 300007-818 300066-288 300066-286 300066-296 300066-298 300087-802 300091-790
Non-Corrosive Gas Flow Valves, Western Enterprises
  • Made from brass to ensure functionality and durability
  • Use these reliable non corrosive gas flow valves with all non-corrosive gases including oxygen
  • Can withstand elevated pressures