Lock Boxes, Brady
Supplier: Safety & Industrial Supplies
262-45190
300065-872EA 104.28 USD
300065-872
Lock Boxes, Brady
Boxes Transport Boxes
These boxes ensures that no one employee has access to the box unless all employees have removed their locks or tags.
  • Accepts up to 13 individual locks
  • Simplifies lockout of large equipment