Dykem® TEXPEN® Industrial Paint Markers
Supplier: Safety & Industrial Supplies
253-16023
300065-694PK 167.98 USD
300065-694
Dykem® TEXPEN® Industrial Paint Markers
Pens Permanent Markers
These oil-based paint markers write sharp, defined, bright marks on any surface including metal, plastic, glass, concrete, rubber, and more. The stainless steel ball tip writes through oil, grease, and water. Paint flow bulb controls paint flow and allows marking at any angle. Rugged aluminum body.
  • Oil-based
  • Bright marks on any surface
  • Stainless steel ball tip writes through oil, grease, or water
  • Paint flow bulb allows marking at any angle
  • Rugged aluminum body
Tip size: 0.24 cm (3/32")