Anchor* High Heat Glove 4in Large
Supplier: ORS NASCO, INC.
Anchor* High Heat Glove, Material: Thermaleather/Wool, Color: Brown, Good abrasion resistance, Lined with heat-resistant 100% wool, Clute Pattern, Cuff Style: Gauntlet, Temperature Range: 400-500 deg F, Cuff Length: 4in, Size: Large