Concrete D-Ring Anchors, DBI-SALA, ORS Nasco
Supplier: Safety & Industrial Supplies
098-2104560
300058-302EA 192.04 USD
300058-302
Concrete D-Ring Anchors, DBI-SALA, ORS Nasco
Fall Protection Fall Arresters
It provides cadmium plated D-ring for connection to fall arrest lanyard or lifeline and special red sheering bolt that falls off to expose a green bolt when proper torque is reached.
  • Removable and partial reusable
  • This simple to use concrete anchor is designed to be set into holes drilled in 3,000 psi concrete