AC Axial Blowers w/Canister, Allegro®
Supplier: Safety & Industrial Supplies
037-9514
300056-836EA 1944.33 USD
300056-836
AC Axial Blowers w/Canister, Allegro®
Ventilators
Complete unit with 15’ (4.57 m) ducting and storage canister provides high output, designed for easy use and storage without sacrificing airflow.
  • Epoxy powder coated construction in “safety orange”
  • Glass reinforced polypropylene (PPG) six blade fan with aluminum hub
  • Design allows ducting to collapse into a built-in canister for easy storage
The unit can also be stored vertically to use a minimum amount of space.
Certifications: Axial Blower is certified to CSA STD C22.2 No 113. Air delivery is certified CEESI.