Fit-Check Banana Oil Ampules, Allegro®
Supplier: Safety & Industrial Supplies
037-0201
300056-772PK 25.47 USD
300056-772
Fit-Check Banana Oil Ampules, Allegro®
Ampules
Provides a fast, safe, and easy method to perform qualitative respirator fit check.
When activated, the ampules give off a non-irritating odor of banana oil (Iso Amyl Acetate). If banana odor is detected by the wearer, the respirator must be adjusted or replaced.