Diaphragm Pumps, Ingersoll-Rand
Supplier: Safety & Industrial Supplies
5011130987
300054-890EA 888.57 USD
300054-890
Diaphragm Pumps, Ingersoll-Rand
Pumps Vacuum Pumps
Pumps feature a patented "unbalanced" air valve design that avoids stall-out, even under low air inlet pressures
  • PTFE diaphragm
  • Polypropylene body
  • For use with all materials, except for highly abrasive substances
Utilize bolted fasteners for leak-tight integrity.