UXT™ Universal "X-tra Tough" Sorbents, SPC
Supplier: Safety & Industrial Supplies
655-UXT200
300054-296CS 163.97 USD
300054-296
UXT™ Universal "X-tra Tough" Sorbents, SPC
Spill Control Materials
  • Abrasion resistant and nonabrasive
  • Highly effective against oils, acids, solvents, bases, chemicals, coolants and antifreeze
  • Non-woven structure on both sides and a low-linting meltblown core
  • Is significantly stronger than ordinary meltblown polypropylene and remains intact even when saturated
  • Full size, heavy weight perfed