Whisper Jet® Safety Air Guns, Guardair
Supplier: Safety & Industrial Supplies
335-80
300052-524EA 73.18 USD
300052-524
Whisper Jet® Safety Air Guns, Guardair
Pneumatic Tools
Safety air guns comes with air volume control.
  • Knurled adjusting nut for air volume control
  • Pistol-style body with hanging hook
  • Venturi nozzle delivers 100% more thrust compared to conventional air guns
  • Cast aluminum body
  • Aluminum Coandaire® nozzle