Jet Guard Safety Air Gun with Tamper-Proof Nozzle, Guardair, Simco Ion
Supplier: Safety & Industrial Supplies
335-74S
300052-506EA 52.41 USD
300052-506
Jet Guard Safety Air Gun with Tamper-Proof Nozzle, Guardair, Simco Ion
Ionizers
Venturi nozzle delivers 100% more thrust compared to conventional air guns
  • Cast aluminum body
  • Trigger control
  • 35 cfm air consumption/flow
  • 0.4 lbs. output
  • 102 dB noise level
  • 1/4" (FNPT) inlet size
Proprietary, protective air cone shields against "chip fly-back." Ideal for close-in applications.
Certifications: Meets OSHA Standards.