Howard Leight Fusion® Multiple-Use Earplugs, Honeywell Safety
Supplier: Safety & Industrial Supplies
154-FUS30S-HP
300052-094PK 302.09 USD
300052-094
Howard Leight Fusion® Multiple-Use Earplugs, Honeywell Safety
Earplugs Reusable Earplugs
These earplugs are made of molded material.
  • FlexiFirm® stem is easy to grasp, ensuring easy insertion into the ear canal
  • SoftFlange™ creates comfortable seal in the ear canal for superior comfort and protection
  • Unique detachable nylon cord system adapts to virtually any application
Certifications: Meets ANSI S.3.19-1974 requirements.