Law OTG® Protective Eyewear, Crews®, ORS Nasco
Supplier: Safety & Industrial Supplies
135-OG112AF 135-OG110AF
300051-960PK 58.78 USD
300051-960 300051-958
Law OTG® Protective Eyewear, Crews®, ORS Nasco
Durable nylon frame with 5 position ratcheting temple design for optimum fit.
  • Non slip TPR temples for comfort and grip
  • Over the glass designed to fit over prescription eyewear
  • 99.9% UV protection
Certifications: Meets or Exceeds ANSI Z87.1+ Standard.