White Food Grade Compound with PTFE, Never-Seez
Supplier: Safety & Industrial Supplies
535-NSWT-14
300051-218EA 113.69 USD
300051-218
White Food Grade Compound with PTFE, Never-Seez
Lubricants
Premium quality lubricant composed entirely of ingredients which meet FDA requirements where incidental food contact is possible.
  • Virtually odorless, colorless, and tasteless multi-purpose lubricant for varied services within food handling plants
  • Use with standard grease guns