Plastic Button Style Blending Discs, Weiler®
Supplier: Safety & Industrial Supplies
804-60134
300049-614CS 97.75 USD
300049-614
Plastic Button Style Blending Discs, Weiler®
Abrasives
  • Cloth-backed, coated abrasive discs for use on right angle air tools
  • Plastic button fastening system
  • Flexible back-up pads give the discs a great deal of conformability