Bobcat Flat Style Flap Discs, Weiler®
Supplier: Safety & Industrial Supplies
804-50936 804-50935 804-50914 804-50933 804-50934 804-50916 804-50913 804-50915
300048-314CS 68.36 USD
300048-314 300049-150 300010-280 300045-570 300046-700 300047-846 300047-960 300048-602
Bobcat Flat Style Flap Discs, Weiler®
  • Used on right angle air die grinders for smooth grinding and enhanced conformability
  • Small size allows access to confined areas