Weiler® Banded Crimped Wire End Brushes, ORS Nasco
Supplier: Safety & Industrial Supplies
804-11114 804-11112 804-11113 804-11103 804-11110
300046-754PK 542.71 USD
300046-754 300047-716 300047-718 300047-714 300048-446
Weiler® Banded Crimped Wire End Brushes, ORS Nasco
Brushes
Crimped brushes are ideal for severe applications to remove rust, paint, oxides, and scale.
  • Banded, solid-end prevents flaring
  • Banded brushes provide shorter trim length for more aggressive brushing action
  • Use for parts that could be damaged by the impact of a knot brush
Use with power drills, drill presses, die grinders, and right-angle air die grinders.