No. 270 Taper Gages, L.S. Starrett
Supplier: Safety & Industrial Supplies
681-51292
300047-690EA 206.01 USD
300047-690
No. 270 Taper Gages, L.S. Starrett
Measuring and Leveling Tools
Steel tools.
  • Very useful tool, especially for bearing work and for gaging slots
  • Accurately tapered throughout its entire length for quick and convenient measuring of slot widths, etc.