Steel Protractors, L.S. Starrett
Supplier: Safety & Industrial Supplies
681-50127
300046-746EA 210.26 USD
300046-746
Steel Protractors, L.S. Starrett
Protractors
Steel protector 6"
  • Double graduations from 0-180° in opposite directions permitting the direct reading of angles and supplementary angles
  • The back of the tool is flat for ease of use
  • The blade can be locked firmly at any angle by the lock nut
  • Satin chrome finish for ease of reading and resistance to rust
  • Rectangle head
For setting bevels, transferring angles, small squaring tasks, checking cutter clearances within certain limits, and other applications.