Weiler® Polyflex Crimped Wire Hollow-End Brush, ORS Nasco
Supplier: Safety & Industrial Supplies
804-35350
300047-454EA 103.44 USD
300047-454 300046-496
Weiler® Polyflex Crimped Wire Hollow-End Brush, ORS Nasco
Brushes
Elastomer gradually wears, exposing consistent short trim for maximum aggression and long life.
Applications include cleaning and finishing recessed areas or I.D.s, die, mold, and tool cleaning and polishing, rubber and plastic flash removal, rust and paint removal, scale and slag removal, I.D. pipe cleaning, carbon cleaning, weld cleaning, deburring, and spot facing. Use with power drills, drill presses, die grinders, and right-angle air die grinders.