Scotch® Vinyl Electrical Tapes 33, ORS Nasco, INC.
Supplier: Safety & Industrial Supplies
500-10174
300046-238EA 25.76 USD
300046-238
Scotch® Vinyl Electrical Tapes 33, ORS Nasco, INC.
Tapes
Polyvinyl chloride (PVC) backing that has a high dielectric strength, is flame-retardent and conformable, and provides excellent mechanical protection with minimum bulk.
  • Pressure sensitive rubber based adhesive
  • Inhibits corrosion of electrical conductor
  • Compatible with solid dielectric cable insulation
It is resistant to abrasion, moisture, alkalies, acid, copper corrosion, varying weather conditions including sunlight.