Weiler® Stem-Mounted Wide Conflex Brush, ORS Nasco
Supplier: Safety & Industrial Supplies
804-17621 804-17620 804-17619 804-17604 804-17602 804-17637 804-17622 804-17623 804-17603 804-17608 804-17601
300046-016EA 57.74 USD
300046-016 300046-302 300046-934 300048-140 300048-136 300010-190 300046-018 300046-020 300048-138 300048-144 300048-460
Weiler® Stem-Mounted Wide Conflex Brush, ORS Nasco
Brushes
Designed for light duty applications. Use to remove rust, paint, oxides, and scale
  • Wide face
  • Covers a broader surface area
Can be used with power drills, die grinders, and right-angle air die grinders.