Weiler® Nylox® Basic Section Wheel Brush,ORS Nasco
Supplier: Safety & Industrial Supplies
804-20600
300045-788CS 165.4 USD
300045-788
Weiler® Nylox® Basic Section Wheel Brush,ORS Nasco
Brushes
Designed for light duty deburring and finishing in non-production applications.
  • Low volume manual deburring
  • General use in a tool room or shop
  • Crimped filament